siRNA Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058206.3 | NP_478113.2 | 1 | 88 | ||
NM_058207.2 | NP_478114.2 | 1 | 88 |
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013744.1 | XP_016869233.1 | 1 | 36 | ||
XM_017013745.1 | XP_016869234.1 | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
