siRNA Details
Gene Symbol: ZNF841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136499.1 | NP_001129971.1 | 7 | 772 | ||
NM_001321349.1 | NP_001308278.1 | 5 | 516 | |||
XM_011526833.2 | XP_011525135.1 | 6 | 664 | |||
XM_017026652.1 | XP_016882141.1 | 3 | 321 | |||
XM_017026653.1 | XP_016882142.1 | 7 | 730 | |||
XM_017026654.1 | XP_016882143.1 | 7 | 716 | |||
XM_017026655.1 | XP_016882144.1 | 3 | 318 | |||
XM_017026656.1 | XP_016882145.1 | 6 | 2367 | |||
XM_017026657.1 | XP_016882146.1 | 6 | 661 | |||
XM_017026658.1 | XP_016882147.1 | 3 | 321 | |||
XM_017026659.1 | XP_016882148.1 | 3 | 321 | |||
XM_017026660.1 | XP_016882149.1 | 3 | 321 | |||
XM_017026661.1 | XP_016882150.1 | 3 | 321 | |||
XM_017026662.1 | XP_016882151.1 | 6 | 2367 | |||
XM_017026663.1 | XP_016882152.1 | 6 | 507 | |||
GenBank mRNA | AK131409.1 | - | 5 | 516 | ||
BC136254.1 | - | 4 | 362 | 9020241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
