siRNA Details
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 1389 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 1373 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 1310 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 1212 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 1026 | |||
NM_153488.4 | NP_705692.1 | 3 | 1092 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 1348 |
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 1310 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 1212 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 1092 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 1026 | |||
NM_005361.3 | NP_005352.1 | 4 | 1387 | |||
NM_175742.2 | NP_786884.1 | 6 | 1373 | |||
NM_175743.2 | NP_786885.1 | 6 | 1389 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 1349 | |||
GenBank mRNA | AK290443.1 | - | 5 | 1310 | ||
BC013098.2 | - | 2 | 996 | 4343986 | ||
BC063681.1 | - | 4 | 1176 | 4403536 | ||
BC108720.1 | - | 4 | 1206 | 6065905 | ||
BC112158.1 | - | 1 | 894 | 8327626 | ||
BC112160.1 | - | 1 | 894 | 8327628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
