siRNA Details
Gene Symbol: GAGE12G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098409.1 | NP_001091879.1 | 5 | 515 |
Gene Symbol: GAGE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098413.3 | NP_001091883.3 | 5 | 512 |
Gene Symbol: GAGE12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098408.1 | NP_001091878.1 | 5 | 515 |
Gene Symbol: GAGE12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127199.1 | NP_001120671.1 | 5 | 526 |
Gene Symbol: GAGE12E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098418.1 | NP_001091888.1 | 5 | 515 |
Gene Symbol: GAGE12F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098405.2 | NP_001091875.1 | 5 | 549 | ||
GenBank mRNA | CF780547.1 | - | 5 | 472 |
Gene Symbol: GAGE12H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098410.1 | NP_001091880.1 | 5 | 515 |
Gene Symbol: GAGE12I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001477.1 | NP_001468.1 | 5 | 453 |
Gene Symbol: GAGE12J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098406.2 | NP_001091876.2 | 5 | 522 | ||
GenBank mRNA | BG186708.1 | - | 2 | 198 | ||
BX108227.1 | - | 5 | 463 |
Gene Symbol: GAGE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098412.2 | NP_001091882.2 | 5 | 494 |
Gene Symbol: GAGE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127212.1 | NP_001120684.1 | 5 | 493 | ||
GenBank mRNA | BG184057.1 | - | 2 | 193 |
Gene Symbol: GAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098411.3 | NP_001091881.1 | 5 | 479 |
Gene Symbol: GAGE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001472.2 | NP_001463.1 | 5 | 479 | ||
GenBank mRNA | U19143.1 | - | 5 | 479 |
Gene Symbol: GAGE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098407.1 | NP_001091877.1 | 5 | 512 |
Gene Symbol: GAGE2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127200.2 | NP_001120672.2 | 5 | 512 | ||
GenBank mRNA | BC018052.1 | - | 5 | 492 | 4812462 |
Gene Symbol: GAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U19144.1 | - | 501 |
Gene Symbol: GAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001474.1 | NP_001465.1 | 5 | 481 | ||
GenBank mRNA | U19145.1 | - | 5 | 481 |
Gene Symbol: GAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001475.1 | NP_001466.1 | 5 | 473 | ||
GenBank mRNA | BC024914.1 | - | 5 | 486 | 4471969 | |
U19146.1 | - | 5 | 473 |
Gene Symbol: GAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001476.1 | NP_001467.1 | 5 | 480 | ||
GenBank mRNA | U19147.1 | - | 5 | 480 |
Gene Symbol: GAGE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF055474.1 | - | 5 | 453 | ||
BC031628.1 | - | 5 | 471 | 5168892 | ||
BC081536.1 | - | 4 | 344 | 30524801 |
Gene Symbol: GAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012196.1 | NP_036328.1 | 5 | 468 | ||
GenBank mRNA | AF055473.1 | - | 5 | 468 |
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029934.1 | XP_016885423.1 | 5 | 1419 | ||
XM_017029935.1 | XP_016885424.1 | 5 | 1416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
