siRNA Details
Gene Symbol: C1QTNF9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014442.2 | NP_001014442.2 | 4 | 353 | ||
NM_001135816.1 | NP_001129288.1 | 4 | 250 | |||
GenBank mRNA | AB113650.1 | - | 1 | 24 | ||
BC015452.1 | - | 4 | 355 | |||
BC073902.1 | - | 4 | 353 | |||
BC132842.1 | - | 2 | 189 | |||
BC132846.1 | - | 2 | 189 | |||
BC142999.1 | - | 4 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
