siRNA Details
Gene Symbol: C1QTNF9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014442.2 | NP_001014442.2 | 3 | 245 | ||
GenBank mRNA | BC015452.1 | - | 3 | 247 | ||
BC073902.1 | - | 3 | 245 | |||
BC132842.1 | - | 1 | 81 | |||
BC132846.1 | - | 1 | 81 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
