siRNA Details
Gene Symbol: NME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000269.2 | NP_000260.1 | 2 | 215 | ||
NM_198175.1 | NP_937818.1 | 3 | 435 | |||
GenBank mRNA | AF487339.1 | - | 3 | 419 | ||
AK291105.1 | - | 2 | 215 | |||
AK309160.1 | - | 3 | 414 | |||
AL360166.1 | - | 2 | 190 | |||
AL360191.1 | - | 2 | 204 | |||
BC000293.2 | - | 2 | 158 | 2819366 | ||
BC018994.2 | - | 2 | 145 | 4126345 | ||
BE727815.1 | - | 2 | 175 | |||
BG715560.1 | - | 2 | 207 | |||
BG757842.1 | - | 3 | 399 | |||
BM479520.1 | - | 150 | ||||
BU600979.1 | - | 2 | 241 | |||
CR542104.1 | - | 1 | 107 | |||
CR542115.1 | - | 1 | 107 | |||
X17620.1 | - | 2 | 191 |
Gene Symbol: NME1-NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018136.2 | NP_001018146.1 | 2 | 238 | ||
NR_037149.1 | 3 | 458 | ||||
GenBank mRNA | AK313448.1 | - | 2 | 195 | ||
BC107894.1 | - | 3 | 360 | 6717551 | ||
BC133029.1 | - | 2 | 141 | 40147051 | ||
BC133031.1 | - | 2 | 141 | 40147053 | ||
BG388883.1 | - | 164 | ||||
BI603164.1 | - | 2 | 220 | |||
DQ109675.1 | - | 2 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
