siRNA Details
Gene Symbol: NME1-NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018136.2 | NP_001018146.1 | 8 | 876 | ||
NR_037149.1 | 9 | 1096 | ||||
GenBank mRNA | AK313448.1 | - | 8 | 833 | ||
BC107894.1 | - | 9 | 998 | 6717551 | ||
BC133029.1 | - | 8 | 779 | 40147051 | ||
BC133031.1 | - | 8 | 779 | 40147053 | ||
BG388883.1 | - | 807 | ||||
DQ109675.1 | - | 8 | 796 |
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018137.2 | NP_001018147.1 | 5 | 651 | ||
NM_001018138.1 | NP_001018148.1 | 4 | 509 | |||
NM_001018139.2 | NP_001018149.1 | 5 | 467 | |||
NM_001198682.1 | NP_001185611.1 | 4 | 517 | |||
NM_002512.3 | NP_002503.1 | 5 | 630 | |||
GenBank mRNA | BC002476.2 | - | 5 | 432 | 3349768 | |
BC095458.1 | - | 5 | 441 | 30524221 | ||
BF034471.1 | - | 5 | 554 | |||
BM556410.1 | - | 5 | 467 | |||
BT007045.1 | - | 4 | 400 | |||
CN267160.1 | - | 4 | 372 | |||
CR541718.1 | - | 4 | 400 | |||
EU794639.1 | - | 5 | 449 | |||
FJ655905.1 | - | 274 | ||||
L16785.1 | - | 5 | 449 | |||
M36981.1 | - | 5 | 472 | |||
X58965.1 | - | 5 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
