siRNA Details
Gene Symbol: RPL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199972.1 | NP_001186901.1 | 2 | 189 | ||
NM_021029.5 | NP_066357.2 | 2 | 189 | |||
GenBank mRNA | AK096711.1 | - | 2 | 68 | ||
AY927233.1 | - | 1 | 52 | |||
BC001781.1 | - | 1 | 75 | 3353669 | ||
BC031015.1 | - | 1 | 67 | 4659712 | ||
BC062219.1 | - | 2 | 100 | 6726961 | ||
BC070204.1 | - | 1 | 64 | 4774945 | ||
BF211310.1 | - | 2 | 120 | |||
BG504049.1 | - | 2 | 68 | |||
BG943625.1 | - | 2 | 256 | |||
BP245950.1 | - | 2 | 189 | |||
CR542198.1 | - | 1 | 35 |
Gene Symbol: RPL36A-HNRNPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199973.1 | NP_001186902.1 | 2 | 189 | ||
NM_001199974.1 | NP_001186903.1 | 2 | 189 | |||
GenBank mRNA | DA530036.1 | - | 2 | 64 | ||
DA998453.1 | - | 2 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
