siRNA Details
Gene Symbol: RPL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021029.5 | NP_066357.2 | 3, 4 | 338 | ||
GenBank mRNA | AY927233.1 | - | 1 | 201 | ||
BC001781.1 | - | 1 | 224 | 3353669 | ||
BC031015.1 | - | 1 | 216 | 4659712 | ||
BC062219.1 | - | 3, 4 | 249 | 6726961 | ||
BC070204.1 | - | 1 | 213 | 4774945 | ||
BF211310.1 | - | 3, 4 | 269 | |||
BG504049.1 | - | 3, 4 | 217 | |||
BG943625.1 | - | 405 | ||||
BP245950.1 | - | 3, 4 | 338 | |||
CR542198.1 | - | 1 | 184 |
Gene Symbol: RPL36A-HNRNPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199973.1 | NP_001186902.1 | 3, 4 | 338 | ||
GenBank mRNA | DA998453.1 | - | 3, 4 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
