siRNA Details
Gene Symbol: C330007P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029951.1 | NP_084227.1 | 5, 6 | 637 | ||
GenBank mRNA | AK021190.1 | - | 5, 6 | 630 | ||
AK032304.1 | - | 5, 6 | 552 | |||
AK034943.1 | - | 5, 6 | 578 | |||
AK035825.1 | - | 5, 6 | 626 | |||
AK036910.1 | - | 5, 6 | 560 | |||
AK044209.1 | - | 5, 6 | 574 | |||
AK084447.1 | - | 4, 5 | 463 | |||
AK086009.1 | - | 2, 3 | 2831 | |||
AK141615.1 | - | 5, 6 | 578 | |||
AK184450.1 | - | 2, 3 | 229 | |||
BC021479.1 | - | 782 | 5344198 | |||
BC051144.1 | - | 5, 6 | 550 | 6588980 | ||
BC085302.1 | - | 5, 6 | 525 |
