siRNA Details
Gene Symbol: Gm13242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 1392 | ||
GenBank mRNA | BC096593.1 | - | 5 | 1404 | 30459933 |
Gene Symbol: Zfp600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 1971 | ||
NM_001177546.2 | NP_001171017.1 | 5 | 2064 | |||
GenBank mRNA | BC108352.1 | - | 5 | 1971 | 30462028 |
