siRNA Details
Gene Symbol: Zglp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103168.1 | NP_001096638.1 | 2 | 395 | ||
GenBank mRNA | DQ286956.1 | - | 2 | 395 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zglp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103168.1 | NP_001096638.1 | 2 | 395 | ||
GenBank mRNA | DQ286956.1 | - | 2 | 395 |