siRNA Details
Gene Symbol: Krtap10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024709.3 | NP_001019880.2 | 1 | 957 | ||
GenBank mRNA | AK079449.1 | - | 1 | 958 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024709.3 | NP_001019880.2 | 1 | 957 | ||
GenBank mRNA | AK079449.1 | - | 1 | 958 |