siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 21 | 3520 | ||
NM_001256198.1 | NP_001243127.1 | 21 | 3520 | |||
NM_172703.3 | NP_766291.2 | 21 | 3424 | |||
GenBank mRNA | AK029440.1 | - | 4 | 504 | ||
BC072600.1 | - | 21 | 3410 | 6410320 | ||
BC079885.1 | - | 21 | 3474 | |||
BC082549.1 | - | 21 | 3484 |
