siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 21 | 3468 | ||
NM_001256198.1 | NP_001243127.1 | 21 | 3468 | |||
NM_172703.3 | NP_766291.2 | 21 | 3372 | |||
GenBank mRNA | AK029440.1 | - | 4 | 452 | ||
BC023898.1 | - | 8 | 1214 | |||
BC024456.1 | - | 9 | 1325 | |||
BC047531.1 | - | 10 | 1539 | |||
BC072600.1 | - | 21 | 3358 | 6410320 | ||
BC075705.1 | - | 6 | 881 | |||
BC079885.1 | - | 21 | 3422 | |||
BC082549.1 | - | 21 | 3432 |

More Information
Other Species Targeted

Gene Ontology Categories:
