siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 5 | 908 | ||
NM_001109686.2 | NP_001103156.1 | 5 | 887 | |||
NM_001109687.2 | NP_001103157.1 | 5 | 908 | |||
NM_172545.4 | NP_766133.2 | 5 | 887 | |||
GenBank mRNA | AB205007.1 | - | 5 | 914 | ||
AK049454.1 | - | 2 | 158 | |||
AK052174.1 | - | 5 | 870 | |||
AK054555.1 | - | 5 | 855 | |||
AK082062.1 | - | 5 | 898 | |||
AK086833.1 | - | 3 | 773 | |||
BC056938.1 | - | 3 | 609 | |||
BC089302.1 | - | 3 | 755 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108572, XM_008761599, XM_008761600, XM_006233618, XM_008761601, and others.
Gene Ontology Categories:
