siRNA Details
Gene Symbol: Sbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145587.2 | NP_663562.1 | 2 | 636 | ||
GenBank mRNA | BC024114.1 | - | 2 | 377 | 5098034 | |
BC025837.1 | - | 2 | 473 | 5135723 | ||
BC031759.1 | - | 2 | 636 | 4194152 |
