siRNA Details
Gene Symbol: Pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026840.3 | NP_081116.3 | 4, 5 | 929 | ||
GenBank mRNA | AK004179.1 | - | 4, 5 | 821 | ||
AK132625.1 | - | 4, 5 | 930 | |||
BC039803.1 | - | 2, 3 | 313 | |||
BC058275.1 | - | 4, 5 | 921 | 6515674 |
