siRNA Details
Gene Symbol: Ssxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198898.2 | NP_942601.1 | 6, 7 | 556 | ||
GenBank mRNA | AY244794.1 | - | 6, 7 | 556 | ||
BC087939.1 | - | 7, 8 | 636 | |||
BC132618.1 | - | 6, 7 | 556 | 40130895 | ||
BC132644.1 | - | 6, 7 | 556 | 40130921 |
