siRNA Details
Gene Symbol: Trim30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175648.2 | NP_783579.1 | 2 | 761 | ||
GenBank mRNA | AK041488.1 | - | 2 | 761 | ||
AK162946.1 | - | 3195 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175648.2 | NP_783579.1 | 2 | 761 | ||
GenBank mRNA | AK041488.1 | - | 2 | 761 | ||
AK162946.1 | - | 3195 |