siRNA Details
Gene Symbol: Olfr159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019476.1 | NP_062349.1 | 1 | 478 | ||
GenBank mRNA | AY043305.1 | - | 1 | 95 | ||
BC113135.1 | - | 1 | 477 | |||
BC134348.1 | - | 1 | 477 |
