siRNA Details
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004173.2 | NP_001004173.1 | 2 | 251 | ||
GenBank mRNA | BC049995.1 | - | 2 | 252 | 6399201 | |
BC147843.1 | - | 2 | 440 | 9087560 | ||
BC147850.1 | - | 2 | 440 | 9087567 | ||
BY234834.1 | - | 2 | 232 | |||
BY297240.1 | - | 2 | 250 |
