siRNA Details
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 10 | 2032 | ||
XM_005245790.3 | XP_005245847.1 | 9 | 2167 | |||
XM_005245791.4 | XP_005245848.1 | 5 | 1300 | |||
XM_017000757.1 | XP_016856246.1 | 10 | 1939 | |||
XM_017000758.1 | XP_016856247.1 | 9 | 1879 | |||
GenBank mRNA | AB020649.1 | - | 10 | 1808 | ||
BC008002.1 | - | 2 | 69 | |||
BC016488.1 | - | 2 | 714 | |||
BC040441.1 | - | 10 | 2035 | |||
BC042103.1 | - | 2 | 362 | |||
BC068599.1 | - | 2 | 387 |
