siRNA Details
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 9, 10 | 1355 | ||
GenBank mRNA | AK076016.1 | - | 9, 10 | 1280 | ||
AK078349.1 | - | 9, 10 | 1268 | |||
AK082892.1 | - | 9, 10 | 1267 | |||
AK132101.1 | - | 9, 10 | 1267 | |||
AK135779.1 | - | 9, 10 | 1269 | |||
AK135869.1 | - | 8, 9 | 964 | |||
AK135875.1 | - | 8, 9 | 964 | |||
AK135956.1 | - | 9, 10 | 1269 | |||
AK136110.1 | - | 9, 10 | 1267 | |||
AK145531.1 | - | 9, 10 | 1266 | |||
AK145651.1 | - | 9, 10 | 1249 | |||
AK146014.1 | - | 9, 10 | 1267 | |||
AK146439.1 | - | 9, 10 | 1326 | |||
AK146904.1 | - | 9, 10 | 1268 | |||
AK146967.1 | - | 9, 10 | 1267 | |||
AK147062.1 | - | 9, 10 | 1326 | |||
AK148852.1 | - | 9, 10 | 1295 | |||
AK153913.1 | - | 9, 10 | 1312 | |||
AK160023.1 | - | 9, 10 | 1267 | |||
AK214411.1 | - | 1, 2 | 18 | |||
AK215897.1 | - | 1, 2 | 18 | |||
AK219296.1 | - | 1, 2 | 18 | |||
BC003785.1 | - | 9, 10 | 1284 | 3600013 | ||
U35142.1 | - | 9, 10 | 1294 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031816, AF090306, BC062012.
Gene Ontology Categories:
