siRNA Details
Gene Symbol: Tmem207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101640.1 | NP_001095110.1 | 2 | 152 | ||
GenBank mRNA | AK142524.1 | - | 2 | 151 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101640.1 | NP_001095110.1 | 2 | 152 | ||
GenBank mRNA | AK142524.1 | - | 2 | 151 |