siRNA Details
Gene Symbol: Gm1673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033458.3 | NP_001028630.1 | 3 | 263 | ||
GenBank mRNA | AK131594.1 | - | 2 | 232 | ||
AK141724.1 | - | 3 | 264 | |||
BC048625.1 | - | 3 | 342 | |||
BC055794.1 | - | 3 | 214 | |||
BC100514.1 | - | 302 | ||||
BC147444.1 | - | 3 | 304 | 9056086 | ||
BC147468.1 | - | 3 | 304 | 9056110 |

More Information
Other Species Targeted

Gene Ontology Categories:
