siRNA Details
Gene Symbol: 1700018F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027069.3 | NP_081345.2 | 5 | 738 | ||
GenBank mRNA | AK006086.1 | - | 5 | 739 | ||
BC147518.1 | - | 5 | 714 | 9056510 | ||
BC147521.1 | - | 5 | 714 | 9056513 |
Gene Symbol: Gm4871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101463.1 | NP_001094933.1 | 5 | 748 |
