siRNA Details
Gene Symbol: Gm2663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102660.1 | NP_001096130.1 | 5 | 721 |
Gene Symbol: 1810009J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023707.3 | NP_076196.1 | 5 | 718 | ||
GenBank mRNA | AK007406.1 | - | 5 | 718 | ||
BC132243.1 | - | 5 | 736 | 40130520 | ||
BC132245.1 | - | 5 | 736 | 40130522 |
