siRNA Details
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136115.2 | NP_001129587.1 | 2 | 387 | ||
NM_015176.3 | NP_055991.1 | 2 | 387 | |||
NR_049764.1 | 2 | 387 | ||||
GenBank mRNA | AB007952.1 | - | 2 | 138 | ||
AK001628.1 | - | 2 | 363 | |||
AK303381.1 | - | 2 | 355 | |||
AK307892.1 | - | 2 | 359 | |||
BC031576.1 | - | 2 | 356 | 5188219 | ||
BP249147.1 | - | 2 | 385 | |||
DB136175.1 | - | 2 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
