siRNA Details
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136115.2 | NP_001129587.1 | 4 | 819 | ||
NM_015176.3 | NP_055991.1 | 5 | 1015 | |||
NR_049764.1 | 4 | 876 | ||||
GenBank mRNA | AB007952.1 | - | 5 | 766 | ||
AK001628.1 | - | 5 | 991 | |||
AK303381.1 | - | 4 | 844 | |||
AK307892.1 | - | 4 | 791 | |||
BC031576.1 | - | 5 | 984 | 5188219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
