siRNA Details
Gene Symbol: BC052040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145898.1 | NP_001139370.1 | 9 | 739 | ||
NM_207264.4 | NP_997147.3 | 9 | 739 | |||
GenBank mRNA | AK090080.1 | - | 9 | 679 | ||
AK154260.1 | - | 9 | 717 | |||
BC052040.1 | - | 9 | 722 | 5718089 |
