siRNA Details
Gene Symbol: Zfp534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127188.2 | NP_001120660.1 | 4 | 890 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127188.2 | NP_001120660.1 | 4 | 890 |