siRNA Details
Gene Symbol: Gm1564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127576.2 | NP_001121048.1 | 5 | 2512 | ||
GenBank mRNA | CJ046726.1 | - | 1 | 273 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm1564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127576.2 | NP_001121048.1 | 5 | 2512 | ||
GenBank mRNA | CJ046726.1 | - | 1 | 273 |