siRNA Details
Gene Symbol: Gm1564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127576.2 | NP_001121048.1 | 8 | 3027 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006247481, XM_220997.
Gene Ontology Categories:
