siRNA Details
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010911.2 | NP_035041.2 | 8, 9 | 1000 | ||
GenBank mRNA | AJ222660.1 | - | 8, 9 | 999 | ||
AK005724.1 | - | 8, 9 | 990 | |||
AK075575.1 | - | 8, 9 | 998 | |||
AK132838.1 | - | 8, 9 | 1001 | |||
AK148527.1 | - | 8, 9 | 1001 | |||
AK159828.1 | - | 8, 9 | 989 | |||
AK183689.1 | - | 406 | ||||
BC010586.1 | - | 8, 9 | 978 | 3711352 |
