siRNA Details
Gene Symbol: Cypt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039943.2 | NP_001035032.1 | 1, 2 | 288 | ||
GenBank mRNA | AM040456.1 | - | 1, 2 | 288 |
Gene Symbol: Cypt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF463502.1 | - | 1, 2 | 275 | ||
AK006892.1 | - | 1, 2 | 297 | |||
AM040454.1 | - | 1, 2 | 288 | |||
AM040455.1 | - | 1, 2 | 288 | |||
BC048455.1 | - | 1, 2 | 293 | |||
BC061079.1 | - | 1, 2 | 293 | 6773161 |
