siRNA Details
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122603.1 | NP_001116075.1 | 15 | 6248 | ||
GenBank mRNA | AK040441.1 | - | 5 | 2952 | ||
BC026653.1 | - | 1 | 453 | |||
BC029250.1 | - | 3 | 1271 | |||
BC030871.1 | - | 4 | 2097 | |||
BC036158.1 | - | 1 | 290 | |||
BC039286.1 | - | 6 | 2674 | |||
BC069956.1 | - | 1 | 256 | |||
BC092067.1 | - | 1 | 327 |
