siRNA Details
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009282.3 | NP_033308.2 | 25 | 2873 | ||
GenBank mRNA | BC062954.1 | - | 25 | 2873 | 30537388 | |
BC069841.1 | - | 26 | 2801 | |||
Z75332.1 | - | 25 | 2922 |

More Information
Other Species Targeted

Gene Ontology Categories:
