siRNA Details
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 36 | 6289 | ||
GenBank mRNA | AK132223.1 | - | 10 | 1414 | ||
AK220176.1 | - | 8 | 1095 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 36 | 6289 | ||
GenBank mRNA | AK132223.1 | - | 10 | 1414 | ||
AK220176.1 | - | 8 | 1095 |