siRNA Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081086.1 | NP_001074555.1 | 14 | 2206 | ||
GenBank mRNA | AK164670.1 | - | 1 | 201 | ||
BC058789.1 | - | 1 | 236 | |||
BC150694.1 | - | 15 | 2227 | 9087605 |

More Information
Other Species Targeted

Gene Ontology Categories:
