siRNA Details
Gene Symbol: 1700034I23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006596.1 | - | 1 | 255 | ||
AK077003.1 | - | 1 | 213 | |||
BC117739.1 | - | 1 | 221 | 40090015 |
Gene Symbol: Gm10731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077188.1 | - | 1 | 206 | ||
BY081654.1 | - | 209 |

More Information
Other Species Targeted

