siRNA Details
Gene Symbol: Nbea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030595.1 | NP_085098.1 | 17, 18 | 2766 | ||
GenBank mRNA | Y18276.1 | - | 17, 18 | 2766 |

More Information
Other Species Targeted

Gene Ontology Categories:
