siRNA Details
Gene Symbol: 9930111J21Rik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173434.1 | NP_775610.1 | 3 | 1501 | ||
GenBank mRNA | AK037088.1 | - | 3 | 1501 | ||
BC119805.1 | - | 2 | 1405 | 40107440 | ||
BC122882.1 | - | 2 | 1410 | 40107441 |
