siRNA Details
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100399.1 | NP_001093869.1 | 8 | 1358 | ||
NM_001100400.1 | NP_001093870.1 | 8 | 1358 | |||
XM_011513672.2 | XP_011511974.1 | 8 | 971 | |||
XM_017007928.1 | XP_016863417.1 | 8 | 933 | |||
GenBank mRNA | AB621803.1 | - | 8 | 1372 | ||
AF294791.1 | - | 8 | 929 | |||
AK098331.1 | - | 8 | 942 | |||
AY550968.1 | - | 7 | 855 | |||
BC041361.1 | - | 8 | 1362 | |||
BC114218.1 | - | 8 | 1054 | 5579832 | ||
BC126225.1 | - | 7 | 768 | 8991941 | ||
CR749425.1 | - | 7 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
