siRNA Details
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100399.1 | NP_001093869.1 | 24, 25 | 3315 | ||
XM_011513672.2 | XP_011511974.1 | 24, 25 | 2928 | |||
XM_017007928.1 | XP_016863417.1 | 24, 25 | 2890 | |||
GenBank mRNA | AB014548.1 | - | 12, 13 | 1319 | ||
AB621803.1 | - | 24, 25 | 3329 | |||
AF294791.1 | - | 24, 25 | 2886 | |||
BC114218.1 | - | 24, 25 | 3011 | 5579832 | ||
BC126225.1 | - | 23, 24 | 2725 | 8991941 | ||
CR749425.1 | - | 23, 24 | 2684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
