siRNA Details
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 17 | 3400 | ||
XM_005255201.3 | XP_005255258.1 | 18 | 3602 | |||
XM_005255202.3 | XP_005255259.1 | 18 | 3548 | |||
XM_005255203.3 | XP_005255260.1 | 17 | 3473 | |||
XM_005255206.3 | XP_005255263.1 | 18 | 3602 | |||
XM_005255208.2 | XP_005255265.1 | 8 | 2300 | |||
XM_006721025.3 | XP_006721088.1 | 16 | 3341 | |||
XM_011545773.2 | XP_011544075.1 | 18 | 3572 | |||
XM_011545774.1 | XP_011544076.1 | 17 | 3518 | |||
XM_011545775.2 | XP_011544077.1 | 17 | 3482 | |||
XM_011545776.2 | XP_011544078.1 | 17 | 3470 | |||
XM_011545777.2 | XP_011544079.1 | 4 | 1733 | |||
XM_017023085.1 | XP_016878574.1 | 16 | 3353 | |||
XM_017023086.1 | XP_016878575.1 | 16 | 3353 | |||
XM_017023087.1 | XP_016878576.1 | 16 | 3287 | |||
XM_017023088.1 | XP_016878577.1 | 13 | 3074 | |||
GenBank mRNA | AB011128.2 | - | 17 | 3398 | ||
BC150276.1 | - | 17 | 3398 | 9007250 |
