siRNA Details
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291222.1 | NP_001278151.1 | 3 | 832 | ||
NM_010626.3 | NP_034756.2 | 3 | 832 | |||
GenBank mRNA | BC141045.1 | - | 3 | 833 | 9055657 | |
BC145222.1 | - | 3 | 833 | 9053773 | ||
BU701630.1 | - | 2 | 480 | |||
CD348088.1 | - | 2 | 437 |
