siRNA Details
Gene Symbol: Cep162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199316.2 | NP_955020.2 | 17 | 2556 | ||
GenBank mRNA | AK135482.1 | - | 17 | 2556 | ||
BC150998.1 | - | 17 | 2361 | 9087911 | ||
BC151006.1 | - | 17 | 2361 | 9087919 | ||
BC151104.1 | - | 17 | 2361 | 9088017 |
