siRNA Details
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024716.1 | NP_001019887.1 | 14 | 2167 | ||
NM_001039156.1 | NP_001034245.1 | 25 | 6934 | |||
NM_138579.4 | NP_613045.3 | 24 | 6796 | |||
GenBank mRNA | AK053194.1 | - | 14 | 2127 | ||
AK085512.1 | - | 14 | 2138 | |||
AK129417.1 | - | 13 | 3592 | |||
AK170390.1 | - | 14 | 2113 | |||
BC003984.1 | - | 12 | 1959 | |||
BC042760.1 | - | 14 | 2106 | |||
BC053329.1 | - | 14 | 2151 | |||
BY761337.1 | - | 2 | 380 | |||
CA322031.1 | - | 4 | 587 | |||
DQ228000.1 | - | 25 | 6934 | |||
DQ228001.2 | - | 24 | 6796 | |||
DQ278602.1 | - | 23 | 6418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
